Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha 2-Macroglobulin Antibody (MM0087-4H44), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212032AF488
Description
alpha 2-Macroglobulin Monoclonal specifically detects alpha 2-Macroglobulin in Human samples. It is validated for Western Blot.Specifications
alpha 2-Macroglobulin | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
A2M | |
Human recombinant alpha 2-Macroglobulin | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
MM0087-4H44 | |
Western Blot | |
alpha-2-M, alpha-2-macroglobulin, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5, CPAMD5DKFZp779B086, FWP007, S863-7 | |
Mouse | |
Protein G purified | |
Neuroscience | |
2 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction