Learn More
Human alpha-2A Adrenergic R/ADRA2A Alexa Fluor™ 350-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier:  R&D Systems FAB10129U100UG
Catalog No. FB10129U100
Add to Cart
This item is not returnable. View return policy
Description
Description
alpha-2A Adrenergic R/ADRA2A Monoclonal specifically detects alpha-2A Adrenergic R/ADRA2A in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
alpha-2A Adrenergic R/ADRA2A
Monoclonal
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human alpha-2A Adrenergic R/ADRA2A in direct ELISAs.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG1
Flow Cytometry
1006614
Flow Cytometry 0.25-1 ug/10^6 cells
a-2A AdrenergicR, ADRA2, ADRA2A, ADRA2Ralpha-2-adrenergic receptor, platelet type, ADRARZNF32, adrenergic, alpha-2A-, receptor, Alpha-2 adrenergic receptor subtype C10, alpha-2A adrenergic receptor, alpha2A AdrenergicR, Alpha-2A adrenoceptor, Alpha-2A adrenoreceptor, Alpha-2AAR, ALPHA2AAR, alpha-2AAR subtype C10, ZNF32
Synthetic peptide containing Human alpha-2A Adrenergic R/ADRA2A
100 μg
150.0
Human
Supernatant
Product Suggestions
Product Suggestions