Learn More
alpha-2A Adrenergic R/ADRA2A Mouse anti-Human, Alexa Fluor 405, Clone: 1006614, R&D Systems
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB10129V100UG
Catalog No. FB10129V100
Add to cart
This item is not returnable. View return policy
Description
Description
alpha-2A Adrenergic R/ADRA2A Monoclonal antibody specifically detects alpha-2A Adrenergic R/ADRA2A in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
alpha-2A Adrenergic R/ADRA2A
1006614
Flow Cytometry 0.25-1 μg/106 cells
a-2A AdrenergicR, ADRA2, ADRA2A, ADRA2Ralpha-2-adrenergic receptor, platelet type, ADRARZNF32, adrenergic, alpha-2A-, receptor, Alpha-2 adrenergic receptor subtype C10, alpha-2A adrenergic receptor, alpha2A AdrenergicR, Alpha-2A adrenoceptor, Alpha-2A adrenoreceptor, Alpha-2AAR, ALPHA2AAR, alpha-2AAR subtype C10, ZNF32
Synthetic peptide containing Human alpha-2A Adrenergic R/ADRA2A
100 μg
Primary
150
Human
Flow Cytometry
Alexa Fluor™ 405
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide
Mouse
IgG1
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human alpha-2A Adrenergic R/ADRA2A in direct ELISAs.