Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-2A Adrenergic R/ADRA2A Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | alpha-2A Adrenergic R/ADRA2A |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB126215
|
Novus Biologicals
NBP310657100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
alpha-2A Adrenergic R/ADRA2A Polyclonal specifically detects alpha-2A Adrenergic R/ADRA2A in Human samples. It is validated for Western Blot.Specifications
alpha-2A Adrenergic R/ADRA2A | |
Western Blot | |
Unconjugated | |
Rabbit | |
GPCR | |
PBS buffer, 2% sucrose | |
150 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
ADRA2, ADRA2Ralpha-2-adrenergic receptor, platelet type, ADRARZNF32, adrenergic, alpha-2A-, receptor, Alpha-2 adrenergic receptor subtype C10, alpha-2A adrenergic receptor, Alpha-2A adrenoceptor, Alpha-2A adrenoreceptor, Alpha-2AAR, ALPHA2AAR, alpha-2AAR subtype C10 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human alpha-2A Adrenergic R/ADRA2A (NP_000672). Peptide sequence RRPERGPRGKGKARASQVKPGDSLPRRGPGATGIGTPAAGPGEERVGAAK | |
Affinity purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title