Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-2C Adrenergic R/ADRA2C Mouse anti-Human, Mouse, Clone: S330A-51, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259377

Catalog No. NBP259377


Add to cart

Description

Description

alpha-2C Adrenergic R/ADRA2C Monoclonal antibody specifically detects alpha-2C Adrenergic R/ADRA2C in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

alpha-2C Adrenergic R/ADRA2C
S330A-51
Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Purified
ADRA2C
Synthetic peptide amino acids 442-462 (QDFRRSFKHILFRRRRRGFRQ, cytoplasmic C-terminus) of human Alpha-2C adrenergic receptor
Protein G purified
RUO
Store at -20C.
Monoclonal
Detects 50kDa or larger (possibly due to dimerization). Does not cross-react with other adrenergic receptors.
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
PBS pH7.4, 50% glycerol with 0.09% Sodium Azide
ADRA2L2alpha-2CAR, ADRA2RL2, ADRARL2, adrenergic, alpha-2C-, receptor, Alpha-2 adrenergic receptor subtype C4, alpha2-AR-C4, alpha-2C adrenergic receptor, Alpha-2C adrenoceptor, Alpha-2C adrenoreceptor, alpha-2C-adrenergic receptor, Alpha-2CAR, ALPHA2CAR
Mouse
IgG2a
100 ug
GPCR, Neuroscience
Primary
152
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only