Learn More
alpha-2C Adrenergic R/ADRA2C Mouse anti-Human, Mouse, HRP, Clone: S330A-51, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259377H
Catalog No. NBP259377H
Add to cart
Description
Description
alpha-2C Adrenergic R/ADRA2C Monoclonal antibody specifically detects alpha-2C Adrenergic R/ADRA2C in Human, Mouse samples. It is validated for Western Blot.
Specifications
Specifications
alpha-2C Adrenergic R/ADRA2C
S330A-51
Western Blot
Purified
ADRA2C
Synthetic peptide amino acids 442-462 (QDFRRSFKHILFRRRRRGFRQ, cytoplasmic C-terminus) of human Alpha-2C adrenergic receptor
Protein G purified
GPCR, Neuroscience
Primary
152
Human, Mouse
Western Blot
HRP
PBS with No Preservative
ADRA2L2alpha-2CAR, ADRA2RL2, ADRARL2, adrenergic, alpha-2C-, receptor, Alpha-2 adrenergic receptor subtype C4, alpha2-AR-C4, alpha-2C adrenergic receptor, Alpha-2C adrenoceptor, Alpha-2C adrenoreceptor, alpha-2C-adrenergic receptor, Alpha-2CAR, ALPHA2CAR
Mouse
IgG2a
100 ul
Store at 4C in the dark.
Monoclonal
Detects 50kDa or larger (possibly due to dimerization). Does not cross-react with other adrenergic receptors.