Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-2C Adrenergic R/ADRA2C Rabbit anti-Human, Mouse, Rat, Non-species specific, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB11039031

 View more versions of this product

Catalog No. NB11039031

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

alpha-2C Adrenergic R/ADRA2C Polyclonal antibody specifically detects alpha-2C Adrenergic R/ADRA2C in Human, Mouse, Rat, Non-species specific samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

alpha-2C Adrenergic R/ADRA2C
1 mg/mL
Western Blot 1:500-1:1000, Immunohistochemistry 1:100-1:300
Affinity Purified
ADRA2C
Synthetic peptide comprising residues 309-324 [GAEGGAGGADGQGAGP] of the human Alpha 2c protein.
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Alpha-2C Adrenergic Receptor
Western Blot, Immunohistochemistry
Unconjugated
PBS with 0.02% Sodium Azide
ADRA2L2alpha-2CAR, ADRA2RL2, ADRARL2, adrenergic, alpha-2C-, receptor, Alpha-2 adrenergic receptor subtype C4, alpha2-AR-C4, alpha-2C adrenergic receptor, Alpha-2C adrenoceptor, Alpha-2C adrenoreceptor, alpha-2C-adrenergic receptor, Alpha-2CAR, ALPHA2CAR
Rabbit
IgG
0.1 mg
GPCR, Neuroscience
Primary
152
Human, Mouse, Rat, Nonspecies specific
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only