Learn More
alpha-2c Adrenergic Receptor Mouse anti-Human, Mouse, Clone: S330A-51, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA527725
Catalog No. PIMA527725
Add to cart
Description
Description
Alpha-2-adrenergic receptors are members of the G protein-coupled receptor superfamily. They include 3 highly homologous subtypes: alpha2A, alpha2B, and alpha2C. These receptors have a critical role in regulating neurotransmitter release from sympathetic nerves and from adrenergic neurons in the central nervous system. The mouse studies revealed that both the alpha2A and alpha2C subtypes were required for normal presynaptic control of transmitter release from sympathetic nerves in the heart and from central noradrenergic neurons. The alpha2A subtype inhibited transmitter release at high stimulation frequencies, whereas the alpha2C subtype modulated neurotransmission at lower levels of nerve activity. This gene encodes the alpha2C subtype, which contains no introns in either its coding or untranslated sequences.
Specifications
Specifications
alpha-2c Adrenergic Receptor
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
P18825, Q01337
Adra 2c, Alpha-2CAR, ADRA2C, ADRA2L2, ADRARL2, Alpha 2CAR, alpha2 AR C4, alpha2 C4, alpha2C, ALPHA2CAR, Alpha-2 adrenergic receptor subtype C4, Alpha-2C adrenoreceptor, Adrenergic alpha 2C receptor, Alpha 2C adrenergic receptor, Alpha 2C adrenergic receptor subtype C4, Alpha 2C adrenoceptor, Alpha 2C adrenoreceptor, Alpha-2-adrenergic receptor, renal type, alpha2cii-adrenergic receptor, Subtype alpha2 C4
Mouse
IgG2a
100 μg
-20°C
Primary
11553, 152
Immunocytochemistry, Immunofluorescence, Western Blot
S330A-51
Unconjugated
ADRA2C
Liquid
ADRA2C
Synthetic peptide amino acids 442-462 (QDFRRSFKHILFRRRRRGFRQ, cytoplasmic C-terminus) of human Alpha-2C adrenergic receptor
Protein G
RUO
Antibody
Monoclonal
Human, Mouse