Learn More
Alpha 2c Adrenergic Receptor Rabbit anti-Human, Rat, Polyclonal, Neuromics™
Rabbit Polyclonal Antibody
Manufacturer:  Neuromics, IncSupplier Diversity Partner RA1906450UL
Encompass
 View more versions of this product
Catalog No. 50-210-0106
Add to cart
Description
Description
Alpha 2c Adrenergic Receptor Polyclonal antibody specifically detects Alpha 2c Adrenergic Receptor in Human, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Specifications
Specifications
Alpha 2c Adrenergic Receptor
Polyclonal
Unconjugated
ADRA2C
Liquid, Purified
Rabbit
IgG
50 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
152, 24175
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
1 mg/mL
PBS with no preservative
P18825, P22086
[a]2C; Adra 2c; ADRA2C; Adra-2c; ADRA2L2; ADRA2RL2; ADRARL2; Adrenergic alpha2C- receptor class I; adrenergic receptor, alpha 2c; adrenergic, alpha-2C-, receptor; Adrenergic, alpha2C-, receptor class I; adrenoceptor alpha 2C; Alpha 2CAR; Alpha-2 adrenergic receptor subtype C4; alpha2 AR C4; alpha2 C4; alpha2-AR-C4; alpha2C; Alpha-2C adrenergic receptor; Alpha-2C adrenoceptor; Alpha-2C adrenoreceptor; alpha2-C4; alpha-2C-adrenergic receptor; ALPHA2CAR; alpha-2CAR; Alpha-2C-AR; subtype alpha2-C4
309-324 GAEGGAGGADGQGAGP
Affinity chromatography
RUO
Antibody
Polyclonal
Human, Rat