Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha 2c Adrenergic Receptor Rabbit anti-Human, Rat, Polyclonal, Neuromics™ Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

$245.52 - $579.50

Specifications

Antigen Alpha 2c Adrenergic Receptor
Concentration 1 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0106 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
RA1906450UL
50 μL
Each for $245.52
Add to cart
 
50-210-0105 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
RA19064150UL
150 μL
Each for $579.50
Add to cart
 
Description

Description

Alpha 2c Adrenergic Receptor Polyclonal antibody specifically detects Alpha 2c Adrenergic Receptor in Human, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Specifications

Specifications

Alpha 2c Adrenergic Receptor
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
Rabbit
Human, Rat
P18825, P22086
152, 24175
IgG
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
ADRA2C
1 mg/mL
Polyclonal
Liquid, Purified
RUO
PBS with no preservative
[a]2C; Adra 2c; ADRA2C; Adra-2c; ADRA2L2; ADRA2RL2; ADRARL2; Adrenergic alpha2C- receptor class I; adrenergic receptor, alpha 2c; adrenergic, alpha-2C-, receptor; Adrenergic, alpha2C-, receptor class I; adrenoceptor alpha 2C; Alpha 2CAR; Alpha-2 adrenergic receptor subtype C4; alpha2 AR C4; alpha2 C4; alpha2-AR-C4; alpha2C; Alpha-2C adrenergic receptor; Alpha-2C adrenoceptor; Alpha-2C adrenoreceptor; alpha2-C4; alpha-2C-adrenergic receptor; ALPHA2CAR; alpha-2CAR; Alpha-2C-AR; subtype alpha2-C4
309-324 GAEGGAGGADGQGAGP
Polyclonal
Affinity chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit