Alfa Aesar™ alpha,3-Dimethylstyrene

Manufacturer: Alfa Aesar™ H50760MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH50760MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1124-20-5
    Molecular Formula C10H12
    Formula Weight 132.21
    MDL Number MFCD08060972
    UN Number UN3295
    Synonym 1-Isopropenyl-3-methylbenzene
    Chemical Name or Material alpha,3-Dimethylstyrene

    Specifications

    Formula Weight 132.21
    UN Number UN3295
    Quantity 250mg
    Odor Characteristic
    Molecular Formula C10H12
    MDL Number MFCD08060972
    CAS 1124-20-5
    Synonym 1-Isopropenyl-3-methylbenzene
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
    Chemical Name or Material alpha,3-Dimethylstyrene

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit