Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha A Crystallin Monoclonal Antibody (1H3.B8), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA527582
Description
Alpha A Crystallin Monoclonal antibody specifically detects Alpha A Crystallin in Bovine, Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
Alpha A Crystallin | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.2 | |
P02470, P02489, P24622, P24623 | |
CRYAA, CRYAA2 | |
Native Human Alpha Crystallin | |
200 μg | |
Primary | |
Bovine, Human, Mouse, Rat | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Western Blot | |
1H3.B8 | |
Unconjugated | |
CRYAA2 | |
[a]A-crystallin; Acry 1; Acry-1; alpha A crystallin; alpha crystallin A chain; alpha-A-crystallin; alpha-crystallin A chain; Alpha-crystallin A(1-151); Alpha-crystallin A(1-156); Alpha-crystallin A(1-157); Alpha-crystallin A(1-162); Alpha-crystallin A(1-163); Alpha-crystallin A(1-165); Alpha-crystallin A(1-168); Alpha-crystallin A(1-172); Alpha-crystallin A2 chain; Alpha-crystallin A2(1-162); Alpha-crystallin A2(1-168); Alpha-crystallin A2(1-172); CRYA 1; Crya1; Crya-1; CRYAA; CRYAA2; Crystallin Alpha 1; crystallin alpha A; crystallin, alpha 1; crystallin, alpha A; Crystallin, alpha polypeptide A; crystallin, alpha-1; CTRCT9; DAcry-1; Heat shock protein beta-4; HSPB 4; HSPB4; human alphaA-crystallin (CRYA1); lens opacity 18; LOC102724652; lop18; small heat shock protein; wu:fc06h04; zgc:92036 | |
Mouse | |
Protein G | |
RUO | |
102724652, 12954, 1409, 24273, 281718 | |
-20°C | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction