Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha Actinin 2, Mouse anti-Human, Alexa Fluor 700, Clone: ACTN2/3293, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279839AF700
Description
Alpha Actinin 2 Monoclonal antibody specifically detects Alpha Actinin 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
Alpha Actinin 2 | |
ACTN2/3293 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
ACTN2 | |
A recombinant fragment (aa557-692) of human Alpha Actinin 2 protein (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Primary | |
Human |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
actinin, alpha 2, alpha-actinin skeletal muscle, Alpha-actinin skeletal muscle isoform 2, alpha-actinin-2, CMD1AA, F-actin cross-linking protein | |
Mouse | |
IgG1 Kappa | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title