Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha Actinin 2 Mouse anti-Human, Clone: ACTN2/3291, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30746520UG
Description
Alpha Actinin 2 Monoclonal specifically detects Alpha Actinin 2 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
Alpha Actinin 2 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
actinin, alpha 2, alpha-actinin skeletal muscle, Alpha-actinin skeletal muscle isoform 2, alpha-actinin-2, CMD1AA, F-actin cross-linking protein | |
A recombinant fragment (aa557-692) of human Alpha Actinin 2 protein(exact sequence is proprietary) | |
20 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin), Peptide Array | |
ACTN2/3291 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
88 | |
Store at 4°C. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title