Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha Actinin 4 Mouse anti-Human, Clone: 93, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Alpha Actinin 4
Clone 93
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122234
View Documents
Novus Biologicals
NBP30738020UG
20 μg
Each for $138.37
Add to cart
 
NB122233
View Documents
Novus Biologicals
NBP30738010UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

Alpha Actinin 4 Monoclonal antibody specifically detects Alpha Actinin 4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Alpha Actinin 4
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
actinin alpha4 isoform, actinin, alpha 4, ACTININ-4, alpha-actinin-4, DKFZp686K23158, F-actin cross-linking protein, focal segmental glomerulosclerosis 1, FSGS, FSGS1, Non-muscle alpha-actinin 4
Recombinant full-length human Alpha Actinin 4 protein
Monoclonal
Protein A or G purified
93
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Phospho Specific
10 mM PBS with 0.05% BSA
81
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit