Learn More
alpha-actinin (Actinin alpha 1) Mouse anti-Human, Clone: OTI7A4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500072

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Alpha Actinin 1 belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it is involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments. This gene encodes a nonmuscle, cytoskeletal, alpha actinin isoform and maps to the same site as the structurally similar erythroid beta spectrin gene. Three transcript variants encoding different isoforms have been found for this gene.Specifications
alpha-actinin (Actinin alpha 1) | |
Monoclonal | |
Unconjugated | |
ACTN1 | |
BDPLT15 | |
Mouse | |
Affinity Chromatography | |
RUO | |
87 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI7A4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P12814 | |
ACTN1 | |
Human recombit protein fragment corresponding to amino acids 650-893 of human alpha-actinin produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.