Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AP2A1 Polyclonal Antibody

alpha Adaptin Polyclonal Antibody
Supplier: Thermo Scientific PA5106390
Description
alpha Adaptin Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| AP2A1 | |
| Polyclonal | |
| Unconjugated | |
| AP2A1 | |
| 100 kDa coated vesicle protein A; adapter-related protein complex 2 alpha-1 subunit; adapter-related protein complex 2 subunit alpha-1; adaptin, alpha A; Adaptor protein complex AP-2 subunit alpha-1; adaptor protein complex AP-2, alpha 1 subunit; adaptor related protein complex 2 alpha 1 subunit; adaptor related protein complex 2, alpha 1 subunit; Adaptor-related protein complex 2 subunit alpha-1; adaptor-related protein complex 2, alpha 1 subunit; adaptor-related protein complex AP-2, alpha 1 subunit; ADTAA; ADTAB; Alpha1-adaptin; alpha2-adaptin; Alpha-adaptin A; alpha-adaptin C; AP2; AP-2 complex subunit alpha-1; AP2A1; AP2-ALPHA; BOS_17797; CLAPA1; CLAPA2; Clathrin assembly protein complex 2 alpha-A large chain; clathrin-associated/assembly/adaptor protein, large, alpha 1; HIP9; HIP-9; HYPJ; Plasma membrane adaptor HA2/AP2 adaptin alpha A subunit | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11771, 160, 308578 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95782, P17426 | |
| AP2A1 | |
| A synthesized peptide derived from human AP2A1(Accession O95782), corresponding to amino acid residues A702-N752. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction