Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha,alpha′,2,4,5,6-Hexachloro-m-xylene 98.0+%, TCI America™
Click to view available options
Quantity:
25g
Specifications
Quantity | 25g |
Melting Point | 139°C |
Color | White-Yellow |
UN Number | 3261 |
Formula Weight | 312.82 |
Physical Form | Crystal-Powder at 20°C |
Percent Purity | ≥98.0% (GC) |
Chemical Name or Material | alpha,alpha′,2,4,5,6-Hexachloro-m-xylene |
Safety and Handling
TSCA : No