Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(alpha,alpha,alpha,2,3,5,6-Heptafluoro-p-tolyl)hydrazine 98.0+%, TCI America™
Click to view available options
Quantity:
5g
Specifications
Quantity | 5g |
Melting Point | 80°C |
Color | Undesignated |
Formula Weight | 248.10 |
Physical Form | Crystal-Powder at 20°C |
Percent Purity | ≥98.0% (GC) |
Chemical Name or Material | (alpha,alpha,alpha,2,3,5,6-Heptafluoro-p-tolyl)hydrazine |
Safety and Handling
TSCA : No