Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha,alpha,alpha',alpha'-Tetrabromo-o-xylene, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
250g
13209-15-9
C8H6Br4
421.752
MFCD00000131
LNAOKZKISWEZNY-UHFFFAOYSA-N
1,2-bis dibromomethyl benzene,alpha,alpha,alpha',alpha'-tetrabromo-o-xylene,benzene, 1,2-bis dibromomethyl,unii-3wg2h68vx4,o-xylylene tetrabromide,alpha,alpha,alpha,alpha-tetrabromo-o-xylene,a,a,a',a'-tetrabromo-o-xylene,o-xylene, alpha,alpha,alpha',alpha'-tetrabromo,alpha,alpha,beta,beta-tetrabromo-o-xylene,acmc-209boi
83234
1,2-bis(dibromomethyl)benzene
C1=CC=C(C(=C1)C(Br)Br)C(Br)Br
13209-15-9
C8H6Br4
421.752
MFCD00000131
LNAOKZKISWEZNY-UHFFFAOYSA-N
1,2-bis dibromomethyl benzene,alpha,alpha,alpha',alpha'-tetrabromo-o-xylene,benzene, 1,2-bis dibromomethyl,unii-3wg2h68vx4,o-xylylene tetrabromide,alpha,alpha,alpha,alpha-tetrabromo-o-xylene,a,a,a',a'-tetrabromo-o-xylene,o-xylene, alpha,alpha,alpha',alpha'-tetrabromo,alpha,alpha,beta,beta-tetrabromo-o-xylene,acmc-209boi
83234
1,2-bis(dibromomethyl)benzene
C1=CC=C(C(=C1)C(Br)Br)C(Br)Br
Quantity 250g
Color Colorless to Yellow
Melting Point 114°C to 118°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6H4(CHBr2)2
Formula Weight 421.75
Physical Form Liquid or Low Melting Solid
Percent Purity ≥96.0% (GC)
Chemical Name or Material α,α,α',α'-Tetrabromo-o-xylene, 97%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Harmful in contact with skin.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 236-176-2

TSCA : TSCA