Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha,alpha,alpha′,alpha′-Tetrachloro-o-xylene 97.0+%, TCI America™

Click to view available options
25G
25641-99-0
C8H6Cl4
243.936
MFCD00000831
UFJYKWQUTDGGPV-UHFFFAOYSA-N
1,2-Bis(dichloromethyl)benzene, o-Xylylene Tetrachloride
117576
1,2-bis(dichloromethyl)benzene
C1=CC=C(C(=C1)C(Cl)Cl)C(Cl)Cl
25641-99-0
C8H6Cl4
243.936
MFCD00000831
UFJYKWQUTDGGPV-UHFFFAOYSA-N
1,2-Bis(dichloromethyl)benzene, o-Xylylene Tetrachloride
117576
1,2-bis(dichloromethyl)benzene
C1=CC=C(C(=C1)C(Cl)Cl)C(Cl)Cl
Melting Point 89°C
Boiling Point 274°C
Color White-Yellow
Quantity 25g
UN Number 1759
Formula Weight 243.94
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (GC)
Chemical Name or Material alpha,alpha,alpha′,alpha′-Tetrachloro-o-xylene

EINECSNumber : (3)-0048

TSCA : Yes