Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ alpha,alpha,alpha-Tribromoquinaldine, 97%
Click to view available options
Quantity:
10g
50g
250g
Specifications
Melting Point | 128°C to 131°C |
Quantity | 250g |
Beilstein | 147638 |
Formula Weight | 379.89 |
Percent Purity | 97% |
Chemical Name or Material | alpha,alpha,alpha-Tribromoquinaldine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-347-1
RTECSNumber : VC3250750
TSCA : Yes
Recommended Storage : Ambient temperatures