Learn More
Medchemexpress LLC Alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid | 77521-29-0 | 99.8% | 186.17 g/mol | C7H10N2O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY100815B10MG
Encompass_Preferred
Alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (RS)-AMPA is a glutamate analogue that functions as a potent, selective agonist of AMPA-type ionotropic glutamate receptors. It is used in neuroscience research to activate AMPA receptors and study excitatory neurotransmission and receptor pharmacology. The compound is supplied as the free base at high purity suitable for laboratory experiments.
  • Potent, selective AMPA receptor agonist.
  • High purity suitable for research (99.8%).
  • Supplied as free base form.
  • Includes CAS number 77521-29-0 for unambiguous identification.
  • Available in small quantities for lab-scale experiments (10 MG).
Catalog No. 50-002-12201