Learn More
Bioassay SystemsSupplier Diversity Partner ALPHA-AMYLASE ASSAY KIT DFS Item
Manufacturer:  Bioassay SystemsSupplier Diversity Partner DAMY100
Assay kit, -amylase activity. Method: OD595nm. Samples: serum, heparinized plasma, saliva, urine etc. Procedure: 10 min. Size: 100 tests. Detection Limit: 10 U/L.
Catalog No. 50-489-199