Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha B-crystallin Mouse anti-Human,Mouse,Bovine,Monkey,Rat, KHL, Clone: aB1788G7G6, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen alpha B-crystallin
Applications ELISA, Immunofluorescence, Immunohistochemistry, Inhibition Assays, Western Blot
Conjugate KHL
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN25521MI
View Documents
MilliporeSigma
MABN2552100UL
100 μL
Each for $372.00
Add to cart
 
MABN25522MI
View Documents
MilliporeSigma
MABN255225UL
25 μL
Each for $147.00
Add to cart
 
Description

Description

alpha B-crystallin Monoclonal antibody specifically detects alpha B-crystallin in Human,Mouse,Bovine,Monkey,Rat samples. It is validated for ELISA, Immunofluorescence, Immunohistochemistry, Inhibition, Western Blotting.
Specifications

Specifications

alpha B-crystallin
KHL
Mouse
Alpha-crystallin B chain;Alpha(B)-crystallin;Heat shock protein beta-5;HspB5;Renal carcinoma antigen NY-REN-27;Rosenthal fiber component
KLH-conjugated linear peptide corresponding to 21 amino acids from the N-terminal half of human Alpha(B)-crystallin.
Primary
Stable for 1 year at -20°C from date of receipt.
ELISA, Immunofluorescence, Immunohistochemistry, Inhibition Assays, Western Blot
Purified
Human, Mouse, Bovine, Monkey, Rat
CRYAB, CRYA2, HSOB5
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit