Learn More
Alfa Aesar™ alpha-Bag Cell Peptide (1-7)
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66473MB
 View more versions of this product
Catalog No. AAJ66473MB
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
922.08
25mg
Ala-Pro-Arg-Leu-Arg-Phe-Tyr
-30° to -10°C
HPLC
Solid
RUO
alpha-Bag Cell Peptide (1-7)