Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Bromocinnamaldehyde, 98%, Acros Organics™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Glass bottle
5443-49-2
C9H7BrO
211.06
WQRWNOKNRHCLHV-TWGQIWQCSA-N
alpha-bromocinnamaldehyde,2-bromo-3-phenylacrylaldehyde,alphabrocine,2-bromo-3-phenyl-2-propenal,bromocinnamal,alpha-bromocinnamic aldehyde,ccris 3924,b 37 van,2-propenal, 2-bromo-3-phenyl,cinnamaldehyde, alpha-bromo
5369403
(Z)-2-bromo-3-phenylprop-2-enal
C1=CC=C(C=C1)C=C(C=O)Br
5443-49-2
C9H7BrO
211.06
WQRWNOKNRHCLHV-TWGQIWQCSA-N
alpha-bromocinnamaldehyde,2-bromo-3-phenylacrylaldehyde,alphabrocine,2-bromo-3-phenyl-2-propenal,bromocinnamal,alpha-bromocinnamic aldehyde,ccris 3924,b 37 van,2-propenal, 2-bromo-3-phenyl,cinnamaldehyde, alpha-bromo
5369403
(Z)-2-bromo-3-phenylprop-2-enal
C1=CC=C(C=C1)C=C(C=O)Br
Quantity 1kg
Melting Point 68.0°C to 72.0°C
Packaging Glass bottle
Color Yellow to Ochre
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula C6H5CH=C(Br)CHO
Beilstein 07,358
Formula Weight 211.06
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material α-Bromocinnamaldehyde
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 226-637-6