Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Bromocinnamaldehyde, 98%, Acros Organics™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Glass bottle
5443-49-2
C9H7BrO
211.058
WQRWNOKNRHCLHV-TWGQIWQCSA-N
alpha-bromocinnamaldehyde,2-bromo-3-phenylacrylaldehyde,alphabrocine,2-bromo-3-phenyl-2-propenal,bromocinnamal,alpha-bromocinnamic aldehyde,ccris 3924,b 37 van,2-propenal, 2-bromo-3-phenyl,cinnamaldehyde, alpha-bromo
5369403
(Z)-2-bromo-3-phenylprop-2-enal
C1=CC=C(C=C1)C=C(C=O)Br
5443-49-2
C9H7BrO
211.058
WQRWNOKNRHCLHV-TWGQIWQCSA-N
alpha-bromocinnamaldehyde,2-bromo-3-phenylacrylaldehyde,alphabrocine,2-bromo-3-phenyl-2-propenal,bromocinnamal,alpha-bromocinnamic aldehyde,ccris 3924,b 37 van,2-propenal, 2-bromo-3-phenyl,cinnamaldehyde, alpha-bromo
5369403
(Z)-2-bromo-3-phenylprop-2-enal
C1=CC=C(C=C1)C=C(C=O)Br
Melting Point 68°C to 72°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color Yellow to Ochre
Quantity 50g
Assay Percent Range 98%
Linear Formula C6H5CH=C(Br)CHO
Formula Weight 211.06
Physical Form Crystalline Powder
Percent Purity ≥97.5% (GC)
Chemical Name or Material alpha-Bromocinnamaldehyde
Show More Show Less