Learn More
Alfa Aesar™ alpha-Conotoxin MI
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66753EXD
 View more versions of this product
Catalog No. AAJ66753EXD
Order in Bulk
Request bulk or custom formats
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
1493.71
0.5mg
Gly-Arg-Cys-Cys-His-Pro-Ala-Cys-Gly-Lys-Asn-Tyr-Ser-Cys-NH2 (Disulfide bridges: Cys3-Cys8, Cys4-Cys14)
2° to 8°C
HPLC
Lyophilized powder
RUO
alpha-Conotoxin MI