Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Cyclodextrin, 98+%, Affymetrix/USB™

Click to view available options
Quantity:
5g
25g
Packaging:
Plastic bottle
10016-20-3
C36H60O30
972.846
MFCD00078207
HFHDHCJBZVLPGP-UHFFFAOYSA-N
cyclohexaamylose,cyclomaltohexose,.alpha.-cyclodextrin,schardinger alpha-dextrin,.alpha.-dextrin,.alpha.-cycloamylose,bdbm4,dextrin, .alpha.-cyclo,.alpha.-schardinger dextrin
320760
C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)O)O
10016-20-3
C36H60O30
972.846
MFCD00078207
HFHDHCJBZVLPGP-UHFFFAOYSA-N
cyclohexaamylose,cyclomaltohexose,.alpha.-cyclodextrin,schardinger alpha-dextrin,.alpha.-dextrin,.alpha.-cycloamylose,bdbm4,dextrin, .alpha.-cyclo,.alpha.-schardinger dextrin
320760
C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)O)O
Quantity 5g
Packaging Plastic bottle
Formula Weight 972.84
Physical Form Crystalline powder
Percent Purity ≥98%
Chemical Name or Material alpha-Cyclodextrin