Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Cyclodextrin, 98%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
500 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
10016-20-3
C36H60O30
972.85
HFHDHCJBZVLPGP-UHFFFAOYNA-N
5,10,15,20,25,30-hexakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontane-31,32,33,34,35,36,37,38,39,40,41,42-dodecol
OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC1C(O)C2O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
10016-20-3
C36H60O30
972.85
HFHDHCJBZVLPGP-UHFFFAOYNA-N
5,10,15,20,25,30-hexakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontane-31,32,33,34,35,36,37,38,39,40,41,42-dodecol
OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC1C(O)C2O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

C1620, 10016-20-3, C36H60O30

Spectrum™ alpha-Cyclodextrin , a polysaccharide, can be used as a dietary fiber. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
CAS Max % 1
Percent Purity 98%
Packaging Amber Glass Bottle
Water 0.1
Quantity 25 g