Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ alpha-Cyclopentylmandelic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
427-49-6
C13H16O3
220.268
MFCD00019296
WFLUEQCOAQCQLP-UHFFFAOYSA-N
alpha-cyclopentylmandelic acid,cyclopentylphenylglycolic acid,cyclopentyl hydroxy phenylacetic acid,cyclopentyl-hydroxy-phenyl-acetic acid,cyclopentyl mandelic acid,alpha-cyclopentyl-dl-mandelic acid,a-cyclo pentyl mandelic acid,alpha-cyclopentylmandelicacid,2-rs-cyclopentyl-2-hydroxy-2-phenylacetic acid alpha-cyclopentylmandelic acid,cyclopentylmandelic acid
98283
2-cyclopentyl-2-hydroxy-2-phenylacetic acid
C1CCC(C1)C(C2=CC=CC=C2)(C(=O)O)O
427-49-6
C13H16O3
220.268
MFCD00019296
WFLUEQCOAQCQLP-UHFFFAOYSA-N
alpha-cyclopentylmandelic acid,cyclopentylphenylglycolic acid,cyclopentyl hydroxy phenylacetic acid,cyclopentyl-hydroxy-phenyl-acetic acid,cyclopentyl mandelic acid,alpha-cyclopentyl-dl-mandelic acid,a-cyclo pentyl mandelic acid,alpha-cyclopentylmandelicacid,2-rs-cyclopentyl-2-hydroxy-2-phenylacetic acid alpha-cyclopentylmandelic acid,cyclopentylmandelic acid
98283
2-cyclopentyl-2-hydroxy-2-phenylacetic acid
C1CCC(C1)C(C2=CC=CC=C2)(C(=O)O)O
Boiling Point 100°C (0.5mmHg)
Melting Point 145°C to 151°C
Quantity 25g
Color Brown to White
Formula Weight 220.26
Physical Form Solid
Percent Purity 97%
Chemical Name or Material alpha-Cyclopentylmandelic acid

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger