Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-D-Glucose-1-phosphate disodium salt tetrahydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
alpha-D-Glucose-1-phosphate disodium salt tetra hydrate is a glucose molecule that can be used in cellular catabolism. It is used as an intermediate in chemical and pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
alpha-D-Glucose-1-phosphate disodium salt tetra hydrate is a glucose molecule that can be used in cellular catabolism. It is used as an intermediate in chemical and pharmaceutical research.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
alpha-D-Glucose-1-phosphate disodium salt tetra hydrate is a glucose molecule that can be used in cellular catabolism. It is used as an intermediate in chemical and pharmaceutical research.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
Specifications
Melting Point | ∼138°C |
Quantity | 5 g |
Solubility Information | Soluble in water. |
Formula Weight | 376.16 (304.10 Anhydrous) |
Percent Purity | 98% |
Chemical Name or Material | alpha-D-Glucose-1-phosphate disodium salt tetrahydrate |
Safety and Handling
Hazard Category | H227 |
Precautionary Statement | P210-P280-P370+P378q-P501c |
EINECSNumber | 260-154-1 |
TSCA | No |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction