Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha Dystroglycan Rat anti-Mouse,Human, Unconjugated, Clone: 3D7, MilliporeSigma™

Rat Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen Alpha Dystroglycan
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN25151MI
View Documents
MilliporeSigma
MABN2515100UG
100 μg
Each for $386.00
Add to cart
 
MABN25152MI
View Documents
MilliporeSigma
MABN251525UG
25 μg
Each for $153.50
Add to cart
 
Description

Description

Alpha Dystroglycan Monoclonal antibody specifically detects Alpha Dystroglycan in Mouse,Human samples. It is validated for Western Blotting.
Specifications

Specifications

Alpha Dystroglycan
Unconjugated
Rat
Alpha-DGP;Alpha-DG
Murine alpha-dystroglycan-Fc-Fusion protein.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Western Blot
Purified
Mouse, Human
Dag1, Dag-1
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit