Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Fetoprotein/AFP Mouse anti-Human, Alexa Fluor™ 405, Clone: 10B8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP223510AF405

Catalog No. NP22310A40

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

alpha-Fetoprotein/AFP Monoclonal antibody specifically detects alpha-Fetoprotein/AFP in Human samples. It is validated for ELISA.
Specifications

Specifications

alpha-Fetoprotein/AFP
10B8
ELISA
Conjugated, Purified
AFP
Human alpha-fetoprotein isolated from amniotic fluid adsorbed onto aluminum hydroxide gel
Protein A or G purified
Cancer, Cancer Stem Cells, Developmental Biology, GPCR, Stem Cell Markers, Tumor Biomarkers
Primary
174.0
Human
ELISA
Alexa Fluor™ 405
50 mM sodium borate with 0.05% sodium azide
Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Clone 10B8 is specific for human AFP. Cross-reactivity to other species or similar molecules have not been determined.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit