Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Fetoprotein/AFP Mouse anti-Human, Canine, Monkey, PE-Cyanine7, Clone: OTI2A9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270103PECY7

Catalog No. N270103PEC7


Add to cart

Description

Description

alpha-Fetoprotein/AFP Monoclonal antibody specifically detects alpha-Fetoprotein/AFP in Human, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

alpha-Fetoprotein/AFP
OTI2A9
Flow Cytometry
Affinity Purified, Conjugated
AFP
Full length recombinant protein of human alpha-Fetoprotein/AFP (NP_001125) produced in HEK293T cells.
Immunogen affinity purified
Cancer, Cancer Stem Cells, Developmental Biology, GPCR, Stem Cell Markers, Tumor Biomarkers
Primary
174.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit