Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Fetoprotein/AFP, Mouse anti-Human,Porcine,Canine,Primate, DyLight 488, Clone: SPM334, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP237754G

Catalog No. NB007006


Add to cart

Description

Description

alpha-Fetoprotein/AFP Monoclonal antibody specifically detects alpha-Fetoprotein/AFP in Human, Porcine, Canine, Primate, Bovine (Negative), Feline (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications

Specifications

alpha-Fetoprotein/AFP
SPM334
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Conjugated, Purified
AFP
Alpha fetoprotein (AFP) purified from serum of a hepatoma patient
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP
Mouse
IgG2a Kappa
0.1 mL
Cancer, Cancer Stem Cells, Developmental Biology, GPCR, Stem Cell Markers, Tumor Biomarkers
Primary
Bovine (Negative), Canine, Feline (Negative), Human, Mouse (Negative), Porcine, Primate, Rat (Negative)
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit