Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-Internexin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 9 publications
Supplier: Novus Biologicals NB300139
Description
alpha-Internexin Polyclonal specifically detects alpha-Internexin in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
alpha-Internexin | |
Polyclonal | |
Western Blot 1:10000 - 1:20000, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:500 - 1:1000, Immunoprecipitation | |
P23565 | |
INA | |
Purified recombinant rat alpha internexin expressed in and purified from E. coli. [UniProt# P23565] | |
Unpurified | |
RUO | |
Primary | |
Human and all other mammalian species tested to date. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Supplied as serum with 5mM Sodium Azide | |
66 kDa neurofilament protein, alpha-Inx, FLJ18662, FLJ57501, internexin neuronal intermediate filament protein, alpha, MGC12702, NEF5, Neurofilament 5, neurofilament 5 (66kD), Neurofilament-66, neurofilament-66, tax-binding protein, NF-66alpha-internexin, TXBP-1 | |
Rabbit | |
66 kDa | |
0.1 mL | |
Cancer, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience | |
9118 | |
Human, Mouse, Rat, Porcine, Bovine, Equine, Rabbit | |
Antisera |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction