Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ alpha-Ketoglutaric acid, Free Acid, >98%, Thermo Scientific™

Click to view available options
Quantity:
100g
500g
1kg
Packaging:
Plastic bottle
328-50-7
C5H6O5
146.098
MFCD00004165
KPGXRSRHYNQIFN-UHFFFAOYSA-N
2-ketoglutaric acid,alpha-ketoglutaric acid,2-oxoglutaric acid,2-oxoglutarate,oxoglutaric acid,alpha-ketoglutarate,2-ketoglutarate,alphaketoglutaric acid,alpha-oxoglutaric acid,2-oxo-1,5-pentanedioic acid
51
CHEBI:30915
2-oxopentanedioic acid
C(CC(=O)O)C(=O)C(=O)O
328-50-7
C5H6O5
146.098
MFCD00004165
KPGXRSRHYNQIFN-UHFFFAOYSA-N
2-ketoglutaric acid,alpha-ketoglutaric acid,2-oxoglutaric acid,2-oxoglutarate,oxoglutaric acid,alpha-ketoglutarate,2-ketoglutarate,alphaketoglutaric acid,alpha-oxoglutaric acid,2-oxo-1,5-pentanedioic acid
51
CHEBI:30915
2-oxopentanedioic acid
C(CC(=O)O)C(=O)C(=O)O
Quantity 500g
Packaging Plastic bottle
Melting Point 113°C to 117°C
Formula Weight 146.10
Physical Form Crystalline Powder
Percent Purity ≥98%
Chemical Name or Material alpha-Ketoglutaric acid