Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ alpha-Mannosidase Assay Kit (Colorimetric)

alpha-Mannosidase Assay Kit (Colorimetric) is used for quantitative determination of alpha-Mannosidase activity using colourmetric methods.

Manufacturer:  Abnova™ KA4566

Catalog No. 89-933-279


Add to cart

Description

Description

  • Suitable Sample: Culture media, plasma, serum, tissue
  • Sample Volume: 10 μL
  • Calibration Range: 1 to 250 U/L
Specifications

Specifications

Gene ID: 4125, 23324, 4123; Gene Name: MAN2B1; Gene Alias; LAMAN, MANB; mannosidase, alpha, class 2B, member 1
1 Kit
Kit
Functional Study
Store the kit at 4°C.
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit