Alfa Aesar™ (+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetonitrile, 98%

Manufacturer: Alfa Aesar™ H2584406

Quantity 5g

 View more versions of this product

Catalog No. AAH2584406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 80866-87-1
    Molecular Formula C10H8F3NO
    Formula Weight 215.18
    MDL Number MFCD00001844
    UN Number UN3276
    Synonym 3,3,3-Trifluoro-2-methoxy-2-phenylpropionitrile
    Chemical Name or Material (+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetonitrile

    Specifications

    Formula Weight 215.18
    Boiling Point 76° to 82°C (12mmHg)
    Quantity 5g
    Molecular Formula C10H8F3NO
    MDL Number MFCD00001844
    Health Hazard 2 GHS H Statement
    H302-H312-H332-H315-H319-H227
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Combustible liquid.
    Flash Point 89°C (152°F)
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
    CAS 80866-87-1
    UN Number UN3276
    Synonym 3,3,3-Trifluoro-2-methoxy-2-phenylpropionitrile
    Health Hazard 1 Warning
    Chemical Name or Material (+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetonitrile
    Assay 98%

    GHS H Statement
    H302-H312-H332-H315-H319-H227
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Combustible liquid.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit