Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 98%

Manufacturer:  Alfa Aesar™ H25845MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH25845MD


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

64312-89-6
252.62
UN3265
(+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
C10H8ClF3O2
MFCD00078209
3,3,3-Trifluoro-2-methoxy-2-phenylpropionyl chloride

Specifications

64312-89-6
252.62
UN3265
213° to 214°C
89°C (192°F)
98%
C10H8ClF3O2
MFCD00078209
3,3,3-Trifluoro-2-methoxy-2-phenylpropionyl chloride
250mg
(+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit