Alfa Aesar™ (+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 98%

Manufacturer: Alfa Aesar™ H2584503

Quantity 1g

 View more versions of this product

Catalog No. AAH2584503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 64312-89-6
    Molecular Formula C10H8ClF3O2
    Formula Weight 252.62
    MDL Number MFCD00078209
    UN Number UN3265
    Synonym 3,3,3-Trifluoro-2-methoxy-2-phenylpropionyl chloride
    Chemical Name or Material (+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride

    Specifications

    Formula Weight 252.62
    Molecular Formula C10H8ClF3O2
    MDL Number MFCD00078209
    UN Number UN3265
    Synonym 3,3,3-Trifluoro-2-methoxy-2-phenylpropionyl chloride
    Boiling Point 213° to 214°C
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Flash Point 89°C (192°F)
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    CAS 64312-89-6
    Health Hazard 1 Danger
    Chemical Name or Material (+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
    Assay 98%

    GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    2° to 8°C

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit