Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ alpha-Methyl-DL-phenylalanine, 98%

Click to view available options
Quantity:
0.25g
1g
5g
1132-26-9
C10H13NO2
179.219
MFCD00010512
HYOWVAAEQCNGLE-UHFFFAOYSA-N
alpha-methyl-dl-phenylalanine,2-amino-2-methyl-3-phenylpropionic acid,alpha-methylphenylalanine,h-dl-me phe-oh,h-alpha-me-dl-phe-oh,+/--2-amino-2-methyl-3-phenylpropionic acid,phenylalanine, alpha-methyl,2-amino-2-methyl-3-phenyl-propanoic acid,h-me-dl-phe-oh,2-methylphenylalanine #
108055
2-amino-2-methyl-3-phenylpropanoic acid
CC(CC1=CC=CC=C1)(C(=O)O)N
1132-26-9
C10H13NO2
179.219
MFCD00010512
HYOWVAAEQCNGLE-UHFFFAOYSA-N
alpha-methyl-dl-phenylalanine,2-amino-2-methyl-3-phenylpropionic acid,alpha-methylphenylalanine,h-dl-me phe-oh,h-alpha-me-dl-phe-oh,+/--2-amino-2-methyl-3-phenylpropionic acid,phenylalanine, alpha-methyl,2-amino-2-methyl-3-phenyl-propanoic acid,h-me-dl-phe-oh,2-methylphenylalanine #
108055
2-amino-2-methyl-3-phenylpropanoic acid
CC(CC1=CC=CC=C1)(C(=O)O)N
Formula Weight 179.22
Percent Purity 98%
Melting Point 293°C to 294°C (decomposition)
Quantity 1g
Chemical Name or Material alpha-Methyl-DL-phenylalanine