Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-Methylacyl-CoA Racemase/AMACR Antibody (OTI5F10), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271749AF700
Description
alpha-Methylacyl-CoA Racemase/AMACR Monoclonal specifically detects alpha-Methylacyl-CoA Racemase/AMACR in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry.Specifications
alpha-Methylacyl-CoA Racemase/AMACR | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
AMACR | |
Full length recombinant protein of human alpha-Methylacyl-CoA Racemase/AMACR (NP_055139) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI5F10 | |
Western Blot, Immunohistochemistry | |
2-methylacyl-CoA racemase, alpha-methylacyl-CoA racemase, CBAS4, EC 5.1.99.4, RACE, RM | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism, Prostate Cancer, Signal Transduction | |
23600.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction