Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-Methylacyl-CoA Racemase/AMACR Rabbit anti-Human, Clone: AMACR/4572R, Novus Biologicals™
Rabbit Monoclonal Antibody
$301.00
Specifications
Antigen | alpha-Methylacyl-CoA Racemase/AMACR |
---|---|
Clone | AMACR/4572R |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122658
|
Novus Biologicals
NBP30759220UG |
20 μg |
Each of 1 for $301.00
|
|
|||||
Description
alpha-Methylacyl-CoA Racemase/AMACR Monoclonal specifically detects alpha-Methylacyl-CoA Racemase/AMACR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
alpha-Methylacyl-CoA Racemase/AMACR | |
0.2 mg/ml | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism, Prostate Cancer, Signal Transduction | |
10 mM PBS with 0.05% BSA | |
23600 | |
IgG | |
Protein A or G purified |
AMACR/4572R | |
Western Blot 1-2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human | |
2-methylacyl-CoA racemase, alpha-methylacyl-CoA racemase, CBAS4, EC 5.1.99.4, RACE, RM | |
Recombinant full-length human alpha-Methylacyl-CoA Racemase/AMACR protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title