Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Methylbenzylamine, 99%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle
618-36-0
C8H11N
121.18
RQEUFEKYXDPUSK-UHFFFAOYNA-N
1-phenylethan-1-amine
CC(N)C1=CC=CC=C1
618-36-0
C8H11N
121.18
RQEUFEKYXDPUSK-UHFFFAOYNA-N
1-phenylethan-1-amine
CC(N)C1=CC=CC=C1

M2815, 618-36-0, C8H11N

Spectrum™ alpha-Methylbenzylamine, 99 Percent , also known as 1-Phenylethylamine, is a monoamine compound whose individual enantiomers of this basic compound are useful for performing chiral resolution of acidic compounds. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Amber Glass Bottle
Quantity 500 mL
CAS Max % 100%
UN Number UN2922
Percent Purity Approx. 99%