Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Methylstyrene, 99%, Acros Organics™

Click to view available options
Quantity:
100g
1kg
2.5kg
Packaging:
Glass bottle
Plastic drum
98-83-9,98-29-3
C9H10
118.18
MFCD00008859
XYLMUPLGERFSHI-UHFFFAOYSA-N
alpha-methylstyrene,2-phenylpropene,2-phenyl-1-propene,isopropenylbenzene,benzene, 1-methylethenyl,2-phenylpropylene,1-methyl-1-phenylethylene,alpha-methylstyrol,as-methylphenylethylene,1-phenyl-1-methylethylene
7407
CHEBI:35060
prop-1-en-2-ylbenzene
CC(=C)C1=CC=CC=C1
98-83-9,98-29-3
C9H10
118.18
MFCD00008859
XYLMUPLGERFSHI-UHFFFAOYSA-N
alpha-methylstyrene,2-phenylpropene,2-phenyl-1-propene,isopropenylbenzene,benzene, 1-methylethenyl,2-phenylpropylene,1-methyl-1-phenylethylene,alpha-methylstyrol,as-methylphenylethylene,1-phenyl-1-methylethylene
7407
CHEBI:35060
prop-1-en-2-ylbenzene
CC(=C)C1=CC=CC=C1
Specific Gravity 0.909
Viscosity 0.94 mPa.s (20°C)
Refractive Index 1.5370 to 1.5390
Melting Point -23.0°C
Quantity 2.5kg
Stabilizer 15ppm tert.-butylcatechol
Color Colorless
Packaging Plastic drum
Density 0.9090g/mL
Infrared Spectrum Authentic
Flash Point 45°C
Boiling Point 165.0°C to 169.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5C(CH3)=CH2
Beilstein 05, 484
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether and benzene, soluble in aromatic hydrocarbons and other organic, solvents
Formula Weight 118.18
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material α-Methylstyrene, Stabilized
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning