Learn More
Alpha Scientific™ DIFF-SAFE™ Blood Dispenser for Blood Smears
Cuts the time of smear-making in half, reducing technician fatigue
Supplier: Alpha Scientific™ DS125
Description
Cuts the time of smear-making in half, reducing technician fatigue
- Compared to the typical procedure involving 15 or more steps when sticks or capillaries are used, the procedure using the DIFF-SAFE dispenser cuts the number of steps by two thirds
- Blunt cannula size is 21G × 0.5 in.
Specifications
Blood dispenser | |
21G x 0.5 in. Blunt needle | |
Blood smears | |
25000 Case |