Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Synuclein Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI2F6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271293JF549

Catalog No. N271293J549


Add to cart

Description

Description

alpha-Synuclein Monoclonal antibody specifically detects alpha-Synuclein in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

alpha-Synuclein
OTI2F6
Immunohistochemistry
Affinity Purified, Conjugated
SNCA
Full length recombinant protein of human alpha-Synuclein (NP_000336) produced in HEK293T cells.
Immunogen affinity purified
Alzheimers Research, Apoptosis, Cell Biology, Cytoskeleton Markers, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Markers
Primary
6622.0
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
Lewy body) 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, synuclein, alpha (non A4 component of amyloid precursor)
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit